Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145088.1 | NP_001138560.1 | 2 | 258 | ||
NM_145647.3 | NP_663622.2 | 2 | 258 | |||
XM_005251102.1 | XP_005251159.1 | 2 | 258 | |||
XM_005251103.1 | XP_005251160.1 | 2 | 258 | |||
XM_005251104.1 | XP_005251161.1 | 2 | 258 | |||
XM_011517378.2 | XP_011515680.1 | 2 | 773 | |||
XM_011517379.2 | XP_011515681.1 | 2 | 258 | |||
XM_017013983.1 | XP_016869472.1 | 2 | 258 | |||
XM_017013984.1 | XP_016869473.1 | 2 | 496 | |||
XM_017013985.1 | XP_016869474.1 | 2 | 369 | |||
XM_017013986.1 | XP_016869475.1 | 2 | 230 | |||
XM_017013987.1 | XP_016869476.1 | 2 | 258 | |||
XM_017013988.1 | XP_016869477.1 | 2 | 209 | |||
XM_017013989.1 | XP_016869478.1 | 2 | 372 | |||
XM_017013990.1 | XP_016869479.1 | 2 | 172 | |||
GenBank mRNA | AK056434.1 | - | 2 | 259 | ||
AK094612.1 | - | 2 | 172 | |||
BC101724.1 | - | 2 | 225 | 8069230 | ||
BC105800.1 | - | 2 | 219 | |||
BC111954.1 | - | 2 | 225 | 8327422 | ||
BC143523.1 | - | 2 | 225 | |||
BC143525.1 | - | 2 | 225 | 9052034 |