Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137554.1 | NP_001131026.1 | 3 | 394 | ||
NM_014060.2 | NP_054779.1 | 3 | 582 | |||
GenBank mRNA | AB034206.1 | - | 3 | 491 | ||
AK294834.1 | - | 3 | 362 | |||
AK311993.1 | - | 3 | 544 | |||
AW957695.1 | - | 3 | 393 | |||
AY364258.1 | - | 2 | 234 | |||
BC001013.2 | - | 3 | 305 | 3344210 | ||
BC095461.1 | - | 514 | 30557789 |