Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF396 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322286.1 | NP_001309215.1 | 2 | 227 | ||
NM_001322290.1 | NP_001309219.1 | 3 | 464 | |||
NM_001322291.1 | NP_001309220.1 | 2 | 227 | |||
NM_145756.2 | NP_665699.1 | 2 | 227 | |||
XM_006722432.3 | XP_006722495.1 | 2 | 955 | |||
XM_017025687.1 | XP_016881176.1 | 3 | 1007 | |||
XM_017025688.1 | XP_016881177.1 | 2 | 842 | |||
XM_017025689.1 | XP_016881178.1 | 2 | 227 | |||
GenBank mRNA | AF492004.3 | - | 2 | 227 | ||
AF533251.2 | - | 2 | 227 | |||
AK055775.1 | - | 2 | 227 | |||
BC130288.1 | - | 2 | 214 | 40146319 | ||
BC136314.1 | - | 2 | 214 | 9020301 |