Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145806.3 | NP_665805.2 | 3, 4 | 482 | ||
NR_130127.1 | 3, 4 | 482 | ||||
GenBank mRNA | AK091711.1 | - | 3, 4 | 430 | ||
AK292380.1 | - | 3, 4 | 428 | |||
BC015139.1 | - | 3, 4 | 472 | |||
BC019897.1 | - | 3, 4 | 452 | 5123355 | ||
HY159562.1 | - | 3, 4 | 458 |
Gene Symbol: ZNF511-PRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EU937518.1 | - | 2, 3 | 254 |