Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDARADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080738.3 | NP_542776.1 | 2 | 267 | ||
NM_145861.2 | NP_665860.2 | 2 | 148 | |||
GenBank mRNA | AK096339.1 | - | 2 | 267 | ||
AK290862.1 | - | 2 | 184 | |||
AK291930.1 | - | 2 | 182 | |||
AK314634.1 | - | 2 | 439 | |||
AY028913.1 | - | 2 | 148 | |||
AY028914.1 | - | 2 | 251 | |||
AY071862.1 | - | 2 | 53 | |||
BC114557.1 | - | 2 | 103 | |||
BC114560.1 | - | 2 | 103 | |||
BC128082.1 | - | 2 | 103 | 40006675 |