Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145260.2 | NP_660303.1 | 3 | 1076 | ||
XM_006711942.3 | XP_006712005.1 | 3 | 1945 | |||
GenBank mRNA | AB082568.1 | - | 2 | 777 | ||
AK074591.1 | - | 3 | 1088 | |||
AK122756.1 | - | 2 | 3904 | |||
BC025712.1 | - | 3 | 1076 | 5227308 |