Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYCTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145262.3 | NP_660305.2 | 4 | 683 | ||
NR_026699.1 | 4 | 685 | ||||
NR_026700.1 | 3 | 267 | ||||
NR_026701.1 | 4 | 683 | ||||
XM_017005730.1 | XP_016861219.1 | 3 | 529 | |||
GenBank mRNA | AF448855.1 | - | 4 | 684 | ||
AK074215.1 | - | 3 | 252 | |||
AY134474.1 | - | 2 | 389 | |||
BC021896.1 | - | 3 | 445 | 3917295 | ||
BC036862.1 | - | 4 | 685 | |||
BC042151.1 | - | 4 | 647 | 4577225 | ||
DQ352863.1 | - | 3 | 587 |