Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297608.1 | NP_001284537.1 | 2 | 529 | ||
NM_145263.3 | NP_660306.1 | 2 | 529 | |||
NR_123728.1 | 2 | 529 | ||||
XM_005265716.1 | XP_005265773.1 | 2 | 444 | |||
XM_005265717.2 | XP_005265774.1 | 2 | 444 | |||
XM_011534369.1 | XP_011532671.1 | 2 | 444 | |||
XM_011534370.1 | XP_011532672.1 | 2 | 444 | |||
XM_017007731.1 | XP_016863220.1 | 1 | 43 | |||
GenBank mRNA | AB465501.1 | - | 2 | 155 | ||
AB465502.1 | - | 2 | 155 | |||
AK057468.1 | - | 2 | 441 | |||
AK304386.1 | - | 2 | 444 | |||
AK314799.1 | - | 2 | 399 | |||
BC025396.1 | - | 2 | 436 | 4821894 | ||
DC404003.1 | - | 2 | 529 |