Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRTOMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145307.4 | NP_001138779.1 | 5 | 1059 | ||
NM_001145308.4 | NP_001138780.1 | 3 | 838 | |||
NM_001145309.3 | NP_001138781.1 | 5 | 1059 | |||
NM_001145310.3 | NP_001138782.1 | 5 | 1059 | |||
NM_001205138.3 | NP_001192067.1 | 4 | 922 | |||
NM_001271471.2 | NP_001258400.1 | 5 | 1059 | |||
NM_001318803.1 | NP_001305732.1 | 5 | 1016 | |||
NM_145309.5 | NP_660352.1 | 5 | 1059 | |||
NR_026886.3 | 6 | 1239 | ||||
NR_134858.1 | 3 | 838 | ||||
XM_006718473.3 | XP_006718536.1 | 5 | 1285 | |||
XM_006718474.3 | XP_006718537.1 | 4 | 1324 | |||
XM_011544847.2 | XP_011543149.1 | 5 | 1513 | |||
XM_011544848.2 | XP_011543150.1 | 6 | 1571 | |||
XM_017017359.1 | XP_016872848.1 | 6 | 1676 | |||
XM_017017360.1 | XP_016872849.1 | 6 | 1528 | |||
GenBank mRNA | AF289588.1 | - | 1 | 41 | ||
AK298997.1 | - | 4 | 441 | |||
AK302772.1 | - | 3 | 332 | |||
AK311690.1 | - | 6 | 709 | |||
AK313150.1 | - | 5 | 743 | |||
AK315892.1 | - | 2 | 137 | |||
BC012855.1 | - | 5 | 531 | 3863323 | ||
BI461207.1 | - | 5 | 502 | |||
DA004114.1 | - | 3 | 392 | |||
EU627066.1 | - | 5 | 531 | |||
EU627067.1 | - | 4 | 481 | |||
EU627068.1 | - | 5 | 531 | |||
EU627069.1 | - | 5 | 531 | |||
EU627070.1 | - | 5 | 531 |