Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPESP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145658.3 | NP_663633.1 | 2 | 372 | ||
GenBank mRNA | AF275321.1 | - | 2 | 276 | ||
AY358983.1 | - | 2 | 355 | |||
BC017998.1 | - | 2 | 346 | 4252566 | ||
BG720873.1 | - | 2 | 368 | |||
BM564110.1 | - | 2 | 371 |