Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256414.1 | NP_001243343.1 | 8 | 1439 | ||
NM_002069.5 | NP_002060.4 | 8 | 1576 | |||
GenBank mRNA | AF055013.1 | - | 8 | 1251 | ||
AK304442.1 | - | 8 | 1439 | |||
AL049933.1 | - | 8 | 1554 | |||
BC026326.1 | - | 8 | 1575 | 4791696 | ||
M17219.1 | - | 8 | 1224 |