Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDIPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286585.1 | NP_001273514.1 | 4, 5 | 790 | ||
NM_001286586.1 | NP_001273515.1 | 5, 6 | 840 | |||
NM_006319.4 | NP_006310.1 | 5, 6 | 925 | |||
XM_005255038.4 | XP_005255095.1 | 3, 4 | 812 | |||
GenBank mRNA | AF014807.1 | - | 5, 6 | 841 | ||
AK094468.1 | - | 4, 5 | 759 | |||
AK097691.1 | - | 5, 6 | 1371 | |||
AK131227.1 | - | 3, 4 | 718 | |||
AK131349.1 | - | 4, 5 | 1274 | |||
AK222676.1 | - | 5, 6 | 804 | |||
AK290632.1 | - | 6, 7 | 699 | |||
AK300805.1 | - | 4, 5 | 706 | |||
AK310988.1 | - | 825 | ||||
AK314544.1 | - | 5, 6 | 856 | |||
BC001444.2 | - | 5, 6 | 761 | 3139134 | ||
BI490060.1 | - | 4, 5 | 752 | |||
BT007301.1 | - | 5, 6 | 494 | |||
BX333193.2 | - | 5, 6 | 796 | |||
CR542016.1 | - | 5, 6 | 494 |