Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166425.1 | NP_001159897.1 | 5, 6 | 651 | ||
NM_001282617.1 | NP_001269546.1 | 5, 6 | 692 | |||
NM_001282618.1 | NP_001269547.1 | 4, 5 | 723 | |||
NM_001282619.1 | NP_001269548.1 | 6, 7 | 1099 | |||
NM_001282620.1 | NP_001269549.1 | 5, 6 | 975 | |||
NM_002070.3 | NP_002061.1 | 5, 6 | 766 | |||
GenBank mRNA | AF493906.1 | - | 5, 6 | 587 | ||
AK055574.1 | - | 4, 5 | 676 | |||
AK096299.1 | - | 5, 6 | 868 | |||
AK096595.1 | - | 5, 6 | 1370 | |||
AK126708.1 | - | 5, 6 | 2403 | |||
AK131062.1 | - | 3, 4 | 1414 | |||
AK302330.1 | - | 5, 6 | 620 | |||
AK304473.1 | - | 5, 6 | 531 | |||
AY677118.1 | - | 5, 6 | 587 | |||
BC012138.1 | - | 5, 6 | 747 | 4562585 | ||
BC014627.2 | - | 5, 6 | 704 | 2963802 | ||
BC016995.1 | - | 6, 7 | 1050 | 4403631 | ||
CR456783.1 | - | 5, 6 | 587 | |||
J03004.1 | - | 5, 6 | 664 | |||
X04828.1 | - | 5, 6 | 710 |