Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319077.1 | NP_001306006.1 | 6 | 897 | ||
NM_001319078.1 | NP_001306007.1 | 5 | 818 | |||
NM_001319079.1 | NP_001306008.1 | 6 | 930 | |||
NM_001319080.1 | NP_001306009.1 | 6 | 929 | |||
NM_001319081.1 | NP_001306010.1 | 5 | 816 | |||
NM_001319082.1 | NP_001306011.1 | 6 | 952 | |||
NM_002131.3 | NP_002122.1 | 6 | 899 | |||
NM_145899.2 | NP_665906.1 | 6 | 932 | |||
NM_145901.2 | NP_665908.1 | 5 | 1099 | |||
NM_145902.2 | NP_665909.1 | 5 | 1066 | |||
NM_145903.2 | NP_665910.1 | 5 | 785 | |||
NM_145905.2 | NP_665912.1 | 5 | 788 | |||
GenBank mRNA | AK096863.1 | - | 7 | 839 | ||
AK301434.1 | - | 5 | 792 | |||
AK308245.1 | - | 5 | 1073 | |||
BC004924.2 | - | 6 | 779 | 3605116 | ||
BC008832.2 | - | 6 | 812 | 4308914 | ||
BC015789.1 | - | 5 | 788 | 4866014 | ||
BC063434.1 | - | 5 | 704 | 5399570 | ||
BC067083.1 | - | 6 | 787 | 4110270 | ||
BC071863.1 | - | 4 | 392 | 4651720 | ||
BC071864.1 | - | 6 | 807 | 6083428 | ||
CX785762.1 | - | 5 | 720 | |||
M23614.1 | - | 5 | 819 | |||
M23615.1 | - | 5 | 786 | |||
M23616.1 | - | 6 | 821 | |||
M23617.1 | - | 5 | 714 | |||
M23618.1 | - | 4 | 621 | |||
M23619.1 | - | 6 | 924 | |||
X14957.1 | - | 6 | 841 | |||
X14958.1 | - | 6 | 808 |