Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147128.3 | NP_667339.1 | 2, 3 | 1626 | ||
GenBank mRNA | AF513707.1 | - | 2, 3 | 1626 | ||
AF527533.1 | - | 2, 3 | 953 | |||
AK025240.1 | - | 1, 2 | 68 | |||
AY054122.1 | - | 3, 4 | 390 |