Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145466.1 | NP_001138938.1 | 4 | 959 | ||
NM_147130.2 | NP_667341.1 | 4 | 904 | |||
XM_006715049.3 | XP_006715112.1 | 5 | 850 | |||
XM_011514459.2 | XP_011512761.1 | 5 | 868 | |||
GenBank mRNA | AF031136.1 | - | 4 | 959 | ||
AF031137.1 | - | 4 | 904 | |||
BC018752.1 | - | 4 | 923 | 4849421 | ||
BC052582.1 | - | 4 | 848 | 5934651 |