Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005751.4 | NP_005742.4 | 4, 5 | 637 | ||
NM_147185.2 | NP_671714.1 | 4, 5 | 637 | |||
XM_017011642.1 | XP_016867131.1 | 4, 5 | 645 | |||
XM_017011643.1 | XP_016867132.1 | 4, 5 | 645 | |||
XM_017011644.1 | XP_016867133.1 | 4, 5 | 645 | |||
XM_017011646.1 | XP_016867135.1 | 4, 5 | 645 | |||
XM_017011647.1 | XP_016867136.1 | 4, 5 | 645 | |||
XM_017011648.1 | XP_016867137.1 | 4, 5 | 645 | |||
XM_017011649.1 | XP_016867138.1 | 4, 5 | 645 | |||
XM_017011650.1 | XP_016867139.1 | 4, 5 | 645 | |||
XM_017011651.1 | XP_016867140.1 | 4, 5 | 645 | |||
XM_017011652.1 | XP_016867141.1 | 4, 5 | 645 | |||
XM_017011653.1 | XP_016867142.1 | 4, 5 | 645 | |||
XM_017011654.1 | XP_016867143.1 | 4, 5 | 645 | |||
XM_017011655.1 | XP_016867144.1 | 4, 5 | 304 | |||
XM_017011656.1 | XP_016867145.1 | 3, 4 | 234 | |||
GenBank mRNA | AB019691.1 | - | 4, 5 | 498 | ||
AF026245.1 | - | 5, 6 | 632 | |||
AF247727.4 | - | 4, 5 | 412 | |||
AJ131693.1 | - | 4, 5 | 634 | |||
BC015533.2 | - | 4, 5 | 586 | |||
CR997756.1 | - | 4, 5 | 637 |