Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TVP23C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145301.2 | NP_660344.2 | 6 | 1192 | ||
GenBank mRNA | BC011952.1 | - | 6 | 1092 | 3860433 |