Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTKN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145307.3 | NP_660350.2 | 12 | 3124 | ||
XM_011539456.2 | XP_011537758.1 | 14 | 3245 | |||
XM_017015842.1 | XP_016871331.1 | 13 | 3147 | |||
GenBank mRNA | AY150309.1 | - | 12 | 2968 | ||
CR749217.1 | - | 1 | 1270 |