Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002095.4 | NP_002086.1 | 5 | 754 | ||
XM_011544509.2 | XP_011542811.1 | 5 | 774 | |||
XM_011544510.1 | XP_011542812.1 | 5 | 774 | |||
XM_017013363.1 | XP_016868852.1 | 5 | 614 | |||
XM_017013364.1 | XP_016868853.1 | 5 | 544 | |||
GenBank mRNA | BC030572.2 | - | 5 | 748 | 3955818 | |
S67861.1 | - | 5 | 691 | |||
X63469.1 | - | 5 | 743 |