Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261427.1 | NP_001248356.1 | 21 | 4798 | ||
NM_001261428.1 | NP_001248357.1 | 22 | 4827 | |||
NM_145693.2 | NP_663731.1 | 20 | 4588 | |||
GenBank mRNA | D80010.1 | - | 20 | 4530 |