Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019106.5 | NP_061979.3 | 7 | 863 | ||
NM_145733.2 | NP_663786.2 | 7 | 863 | |||
XM_017028862.1 | XP_016884351.1 | 8 | 2461 | |||
GenBank mRNA | AB209152.1 | - | 4 | 330 | ||
AF285107.1 | - | 7 | 740 | |||
AF285108.1 | - | 5 | 445 | |||
AF285109.1 | - | 7 | 740 | |||
AK290037.1 | - | 7 | 848 | |||
AK299919.1 | - | 6 | 608 | |||
AL833942.1 | - | 7 | 798 | |||
BC111779.1 | - | 7 | 756 | 40031828 | ||
CR456572.1 | - | 7 | 752 |