Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022139.3 | NP_071422.1 | 6 | 1114 | ||
NM_145762.2 | NP_665705.1 | 5 | 1204 | |||
GenBank mRNA | AF253318.1 | - | 6 | 1114 |