Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145913.3 | NP_666018.3 | 13, 14 | 2031 | ||
GenBank mRNA | AF536216.1 | - | 13, 14 | 2004 | ||
AK075263.1 | - | 13, 14 | 1689 | |||
AK313788.1 | - | 13, 14 | 2000 | |||
AY081220.1 | - | 13, 14 | 2005 | |||
BC110492.2 | - | 13, 14 | 1666 | 40007567 |