Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TICRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308025.1 | NP_001294954.1 | 20 | 4183 | ||
NM_152259.3 | NP_689472.3 | 20 | 4186 | |||
GenBank mRNA | AK074727.1 | - | 1 | 678 | ||
AK123612.1 | - | 13 | 2286 | |||
AK315750.1 | - | 1 | 498 | |||
BC033209.1 | - | 1 | 228 | |||
BX538335.1 | - | 7 | 1487 | |||
GQ227787.1 | - | 20 | 4183 |