Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270488.1 | NP_001257417.1 | 4 | 603 | ||
NM_005643.3 | NP_005634.1 | 5 | 700 | |||
XM_011514827.2 | XP_011513129.1 | 5 | 1090 | |||
GenBank mRNA | AF118094.1 | - | 5 | 440 | ||
AK302226.1 | - | 4 | 574 | |||
AK313475.1 | - | 5 | 641 | |||
BC021972.2 | - | 5 | 656 | 3935882 | ||
BT019404.1 | - | 5 | 586 | |||
BT019405.1 | - | 5 | 586 | |||
BX538195.1 | - | 2 | 1968 | |||
BX647568.1 | - | 3 | 2871 | |||
CR542068.1 | - | 5 | 586 | |||
D63705.1 | - | 5 | 669 | |||
X83928.1 | - | 5 | 678 |