Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL14EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152316.2 | NP_689529.1 | 2 | 382 | ||
XM_005252792.3 | XP_005252849.1 | 2 | 407 | |||
GenBank mRNA | AK096287.1 | - | 2 | 379 | ||
AK127509.1 | - | 1 | 3128 | |||
BC047775.1 | - | 2 | 370 | 6062049 | ||
BP341330.1 | - | 2 | 384 | |||
BX647617.1 | - | 2 | 374 |