Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152329.3 | NP_689542.2 | 2 | 520 | ||
NM_203467.1 | NP_982292.1 | 2 | 520 | |||
NR_037792.1 | 2 | 520 | ||||
NR_037793.1 | 3 | 628 | ||||
GenBank mRNA | AK293156.1 | - | 2 | 239 | ||
AY052405.1 | - | 2 | 206 | |||
BC030142.2 | - | 2 | 456 | |||
BC064133.1 | - | 3 | 373 | |||
BC093697.1 | - | 2 | 222 | |||
BC107888.1 | - | 2 | 269 | |||
BC112241.1 | - | 2 | 222 | |||
BC139921.1 | - | 2 | 260 | 40148567 | ||
BI825814.1 | - | 2 | 447 | |||
BI826606.1 | - | 2 | 521 | |||
BI829295.1 | - | 2 | 457 | |||
BQ059807.1 | - | 2 | 190 | |||
BX248298.1 | - | 2 | 263 |