Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042481.2 | NP_001035946.1 | 4 | 673 | ||
NM_001267046.1 | NP_001253975.1 | 3 | 449 | |||
NM_152330.3 | NP_689543.1 | 3 | 449 | |||
XM_006720029.1 | XP_006720092.1 | 3 | 546 | |||
XM_006720030.1 | XP_006720093.1 | 3 | 546 | |||
XM_011536423.1 | XP_011534725.1 | 4 | 693 | |||
XM_011536424.1 | XP_011534726.1 | 5 | 818 | |||
XM_017020967.1 | XP_016876456.1 | 3 | 399 | |||
XM_017020968.1 | XP_016876457.1 | 3 | 399 | |||
GenBank mRNA | AK055545.1 | - | 3 | 360 | ||
AK126521.1 | - | 1 | 94 | |||
AK315764.1 | - | 3 | 370 | |||
AL833158.1 | - | 4 | 659 | |||
BC020521.2 | - | 3 | 397 | 3914276 | ||
BX161430.1 | - | 3 | 449 |