Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM110B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147189.2 | NP_671722.1 | 5 | 1781 | ||
XM_005251324.2 | XP_005251381.1 | 4 | 1775 | |||
XM_005251325.3 | XP_005251382.1 | 3 | 11938 | |||
XM_005251326.2 | XP_005251383.1 | 3 | 11963 | |||
XM_017013948.1 | XP_016869437.1 | 4 | 4350 | |||
GenBank mRNA | AK026141.1 | - | 2 | 1315 | ||
AK092221.1 | - | 5 | 1781 | |||
AY937246.1 | - | 1 | 1013 | |||
BC024294.1 | - | 2 | 1293 | 5440447 | ||
DQ431182.1 | - | 1 | 901 | |||
U79298.1 | - | 1 | 912 |