Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147191.1 | NP_671724.1 | 4 | 895 | ||
GenBank mRNA | AF331526.1 | - | 4 | 895 | ||
AF520613.1 | - | 4 | 895 |