Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147193.2 | NP_671726.2 | 5, 6 | 1631 | ||
XM_017000408.1 | XP_016855897.1 | 6, 7 | 1698 | |||
XM_017000410.1 | XP_016855899.1 | 6, 7 | 1812 | |||
XM_017000411.1 | XP_016855900.1 | 6, 7 | 1698 | |||
GenBank mRNA | AK090634.1 | - | 5, 6 | 1630 | ||
AK093474.1 | - | 3, 4 | 1000 | |||
BC101799.1 | - | 3, 4 | 1137 | 8069305 | ||
BC104911.1 | - | 3, 4 | 1137 | 8143914 |