Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004204.3 | NP_004195.2 | 2 | 551 | ||
NM_148920.2 | NP_683721.1 | 2 | 551 | |||
GenBank mRNA | AB003723.1 | - | 2 | 515 | ||
AF030177.1 | - | 2 | 484 | |||
AK300297.1 | - | 2 | 518 | |||
AK314361.1 | - | 3 | 580 | |||
BC006377.2 | - | 2 | 501 | 4299132 | ||
DA206420.1 | - | 2 | 551 |
Gene Symbol: NHLRC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA560515.1 | - | 3 | 571 |