Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321312.1 | NP_001308241.1 | 8 | 2690 | ||
NM_001321313.1 | NP_001308242.1 | 8 | 2476 | |||
NM_001321314.1 | NP_001308243.1 | 7 | 2396 | |||
NM_001321315.1 | NP_001308244.1 | 6 | 2309 | |||
NM_005966.3 | NP_005957.2 | 10 | 2952 | |||
XM_005246582.1 | XP_005246639.1 | 9 | 2625 | |||
XM_005246583.1 | XP_005246640.1 | 9 | 2739 | |||
XM_005246586.1 | XP_005246643.1 | 8 | 2429 | |||
XM_005246587.1 | XP_005246644.1 | 7 | 2342 | |||
XM_011511219.2 | XP_011509521.1 | 9 | 2594 | |||
XM_017004170.1 | XP_016859659.1 | 9 | 2635 | |||
XM_017004171.1 | XP_016859660.1 | 8 | 2802 | |||
XM_017004172.1 | XP_016859661.1 | 8 | 2574 | |||
XM_017004173.1 | XP_016859662.1 | 9 | 2657 | |||
XM_017004174.1 | XP_016859663.1 | 8 | 2473 | |||
XM_017004175.1 | XP_016859664.1 | 7 | 2551 | |||
XM_017004176.1 | XP_016859665.1 | 8 | 2544 | |||
GenBank mRNA | AF045451.1 | - | 7 | 2399 | ||
AK296549.1 | - | 6 | 1637 |