Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135825.1 | NP_001129297.1 | 4 | 413 | ||
NM_152267.3 | NP_689480.2 | 6 | 581 | |||
NR_024209.1 | 6 | 566 | ||||
NR_024210.1 | 6 | 748 | ||||
NR_024211.1 | 5 | 468 | ||||
NR_024212.1 | 5 | 357 | ||||
GenBank mRNA | AK027035.1 | - | 5 | 280 | ||
AK095947.1 | - | 4 | 322 | |||
AK291236.1 | - | 6 | 490 | |||
AK297414.1 | - | 5 | 438 | |||
BC009504.2 | - | 6 | 530 | 2964524 | ||
BC012817.1 | - | 6 | 539 | 2964524 | ||
BC026040.1 | - | 6 | 473 | 4866294 | ||
BC033166.1 | - | 6 | 516 | 4549320 | ||
BC035684.1 | - | 6 | 540 | 5750180 | ||
BX648019.1 | - | 6 | 490 | |||
CR456349.1 | - | 6 | 469 | |||
DA498404.1 | - | 4 | 386 | |||
DQ296559.1 | - | 6 | 522 | |||
DQ296561.1 | - | 6 | 522 | |||
DQ296562.1 | - | 4 | 354 | |||
DQ296564.1 | - | 6 | 690 | |||
DQ296565.1 | - | 6 | 522 |