Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL14EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152316.2 | NP_689529.1 | 2, 3 | 585 | ||
GenBank mRNA | AK096287.1 | - | 2, 3 | 582 | ||
AK127509.1 | - | 1, 2 | 3331 | |||
BC047775.1 | - | 2, 3 | 573 | 6062049 | ||
BX647617.1 | - | 2, 3 | 578 |