Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFRKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143835.1 | NP_001137307.1 | 10 | 1142 | ||
NM_006165.3 | NP_006156.2 | 8 | 1137 | |||
XM_005271567.3 | XP_005271624.1 | 10 | 1178 | |||
XM_005271568.3 | XP_005271625.1 | 9 | 1093 | |||
XM_011542850.2 | XP_011541152.1 | 11 | 1215 | |||
XM_011542851.2 | XP_011541153.1 | 10 | 1197 | |||
XM_011542852.2 | XP_011541154.1 | 9 | 1035 | |||
XM_017017793.1 | XP_016873282.1 | 9 | 1089 | |||
XM_017017794.1 | XP_016873283.1 | 9 | 1053 | |||
XM_017017795.1 | XP_016873284.1 | 11 | 1179 | |||
XM_017017796.1 | XP_016873285.1 | 9 | 1057 | |||
XM_017017797.1 | XP_016873286.1 | 9 | 1089 | |||
XM_017017798.1 | XP_016873287.1 | 10 | 1142 | |||
XM_017017799.1 | XP_016873288.1 | 11 | 1289 | |||
XM_017017800.1 | XP_016873289.1 | 11 | 1229 | |||
GenBank mRNA | BC063280.1 | - | 10 | 1088 | 5271534 | |
X80878.1 | - | 8 | 1146 |