Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS6ST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077188.1 | NP_001070656.1 | 6 | 2970 | ||
NM_147175.3 | NP_671704.3 | 4 | 2850 | |||
XM_005262490.3 | XP_005262547.1 | 5 | 2870 | |||
XM_005262491.2 | XP_005262548.1 | 4 | 2783 | |||
XM_011531406.1 | XP_011529708.1 | 5 | 2199 | |||
XM_017029944.1 | XP_016885433.1 | 3 | 2750 | |||
XM_017029945.1 | XP_016885434.1 | 5 | 2144 | |||
XM_017029946.1 | XP_016885435.1 | 4 | 2108 | |||
GenBank mRNA | AK027720.1 | - | 3 | 2126 | ||
AK056706.1 | - | 4 | 2850 | |||
AK075402.1 | - | 3 | 2029 | |||
AK295118.1 | - | 4 | 2110 | |||
AL831923.1 | - | 1 | 1042 | |||
BC037325.2 | - | 4 | 2029 | |||
BC094718.1 | - | 3 | 2089 | |||
BC110620.1 | - | 3 | 2689 | 40035499 | ||
BC110621.1 | - | 3 | 2689 | 40035506 |