Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147192.2 | NP_671725.1 | 4 | 2413 | ||
NM_172225.1 | NP_757379.1 | 4 | 2398 | |||
XM_011540668.2 | XP_011538970.1 | 5 | 2644 | |||
XM_017000289.1 | XP_016855778.1 | 5 | 2436 | |||
GenBank mRNA | AB037699.1 | - | 4 | 2398 |