Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004204.3 | NP_004195.2 | 11 | 2479 | ||
NM_148920.2 | NP_683721.1 | 10 | 2417 | |||
GenBank mRNA | AB003723.1 | - | 11 | 2443 | ||
AF030177.1 | - | 11 | 2412 | |||
AK055163.1 | - | 4 | 1720 | |||
AK092073.1 | - | 2 | 1304 | |||
BC006377.2 | - | 10 | 2367 | 4299132 |