Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006496.3 | NP_006487.1 | 8 | 1212 | ||
GenBank mRNA | AF493907.1 | - | 8 | 1055 | ||
AK130069.1 | - | 2 | 208 | |||
AK311691.1 | - | 9 | 1075 | |||
AK312252.1 | - | 8 | 1141 | |||
BC025285.1 | - | 8 | 1165 | 5433334 | ||
J03005.1 | - | 8 | 1063 | |||
J03198.1 | - | 8 | 1117 | |||
J03238.1 | - | 8 | 1145 | |||
M27543.1 | - | 8 | 1097 |