Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135825.1 | NP_001129297.1 | 5 | 743 | ||
NM_152267.3 | NP_689480.2 | 7 | 911 | |||
NR_024209.1 | 7 | 896 | ||||
NR_024210.1 | 7 | 1078 | ||||
NR_024211.1 | 6 | 798 | ||||
NR_024212.1 | 6 | 687 | ||||
GenBank mRNA | AK027035.1 | - | 6 | 609 | ||
AK095947.1 | - | 5 | 652 | |||
AK291236.1 | - | 7 | 820 | |||
AK297414.1 | - | 6 | 768 | |||
BC026040.1 | - | 7 | 803 | 4866294 | ||
BC033166.1 | - | 7 | 846 | 4549320 | ||
BC035684.1 | - | 7 | 870 | 5750180 | ||
BX648019.1 | - | 7 | 820 | |||
DQ296559.1 | - | 7 | 852 | |||
DQ296561.1 | - | 7 | 852 | |||
DQ296562.1 | - | 5 | 684 | |||
DQ296564.1 | - | 7 | 1020 | |||
DQ296565.1 | - | 7 | 852 |