Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152271.3 | NP_689484.3 | 10 | 2028 | ||
XM_005273685.3 | XP_005273742.1 | 10 | 1997 | |||
XM_011544694.2 | XP_011542996.1 | 10 | 1780 | |||
GenBank mRNA | AK074329.1 | - | 5 | 580 | ||
AK297266.1 | - | 9 | 1165 | |||
AK300538.1 | - | 6 | 774 | |||
BC004538.2 | - | 2 | 164 | |||
BC106042.1 | - | 10 | 1294 | |||
BC117381.1 | - | 7 | 959 | 40125932 | ||
BC117385.1 | - | 7 | 959 | 40125936 | ||
BQ718281.1 | - | 625 |