Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145006.1 | NP_001138478.1 | 4 | 2006 | ||
NM_001145007.1 | NP_001138479.1 | 4 | 1945 | |||
NM_152291.2 | NP_689504.2 | 3 | 1908 | |||
GenBank mRNA | AK093828.1 | - | 5 | 1485 | ||
BC025688.1 | - | 3 | 1806 | 5205526 | ||
BX537739.1 | - | 1 | 1981 | |||
BX647190.1 | - | 5 | 1641 | |||
BX648903.1 | - | 2 | 1357 | |||
BX648904.1 | - | 2 | 1490 | |||
L13283.1 | - | 3 | 1815 |