Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152311.3 | NP_689524.1 | 3 | 833 | ||
XM_011539274.2 | XP_011537576.1 | 2 | 652 | |||
GenBank mRNA | BC029478.1 | - | 3 | 833 | 4733535 | |
BM830163.1 | - | 3 | 586 | |||
BX640823.1 | - | 2 | 1113 |