Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113178.2 | NP_001106649.1 | 8 | 1773 | ||
NM_006599.3 | NP_006590.1 | 7 | 1719 | |||
NM_138713.3 | NP_619727.2 | 8 | 1773 | |||
NM_138714.3 | NP_619728.2 | 9 | 1835 | |||
NM_173214.2 | NP_775321.1 | 8 | 1781 | |||
NM_173215.2 | NP_775322.1 | 8 | 1781 | |||
XM_006721125.3 | XP_006721188.1 | 7 | 2444 | |||
XM_011522817.2 | XP_011521119.1 | 7 | 1719 | |||
XM_011522818.2 | XP_011521120.1 | 10 | 1908 | |||
XM_011522819.2 | XP_011521121.1 | 10 | 1490 | |||
XM_011522820.2 | XP_011521122.1 | 7 | 920 | |||
XM_011522822.2 | XP_011521124.1 | 6 | 868 | |||
XM_017022870.1 | XP_016878359.1 | 7 | 2068 | |||
XM_017022871.1 | XP_016878360.1 | 8 | 1795 | |||
XM_017022872.1 | XP_016878361.1 | 7 | 1649 | |||
XM_017022873.1 | XP_016878362.1 | 7 | 1261 | |||
GenBank mRNA | AB020634.1 | - | 7 | 1701 | ||
AF089824.1 | - | 8 | 1723 | |||
AF134870.1 | - | 5 | 1155 | |||
AF163836.1 | - | 6 | 1242 | |||
AF346509.1 | - | 9 | 2721 | |||
AJ005683.1 | - | 8 | 1397 | |||
AJ005684.1 | - | 8 | 1397 | |||
AJ243298.1 | - | 8 | 1540 | |||
AJ243299.1 | - | 8 | 1532 | |||
BC131509.1 | - | 8 | 1677 | 40083218 | ||
BC146765.1 | - | 7 | 1701 | 9007208 |