Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172681.1 | NP_001166152.1 | 6 | 1983 | ||
NM_001172682.1 | NP_001166153.1 | 5 | 1889 | |||
NM_152320.2 | NP_689533.2 | 7 | 2156 | |||
XM_005268637.3 | XP_005268694.1 | 5 | 1936 | |||
XM_005268638.3 | XP_005268695.1 | 6 | 2176 | |||
XM_005268639.2 | XP_005268696.1 | 5 | 1827 | |||
XM_011537897.1 | XP_011536199.1 | 6 | 2156 | |||
XM_011537898.1 | XP_011536200.1 | 6 | 2084 | |||
XM_017018797.1 | XP_016874286.1 | 5 | 1980 | |||
XM_017018798.1 | XP_016874287.1 | 6 | 2010 | |||
GenBank mRNA | AK055857.1 | - | 6 | 2059 | ||
AK092740.1 | - | 6 | 2583 | |||
AL713659.1 | - | 1 | 1041 | |||
AY842285.1 | - | 6 | 2610 |