Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152325.1 | NP_689538.1 | 4 | 438 | ||
XM_011534919.2 | XP_011533221.1 | 4 | 498 | |||
XM_011534921.2 | XP_011533223.1 | 4 | 502 | |||
XM_011534924.2 | XP_011533226.1 | 4 | 502 | |||
XM_011534925.2 | XP_011533227.1 | 4 | 502 | |||
XM_011534926.2 | XP_011533228.1 | 4 | 502 | |||
XM_011534927.2 | XP_011533229.1 | 4 | 502 | |||
GenBank mRNA | BC030277.1 | - | 4 | 438 | 5167551 |