Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042481.2 | NP_001035946.1 | 15 | 4289 | ||
NM_001267046.1 | NP_001253975.1 | 14 | 4089 | |||
NM_001267047.1 | NP_001253976.1 | 5 | 2846 | |||
NM_152330.3 | NP_689543.1 | 14 | 4065 | |||
XM_006720029.1 | XP_006720092.1 | 14 | 4186 | |||
XM_006720030.1 | XP_006720093.1 | 14 | 4162 | |||
XM_011536423.1 | XP_011534725.1 | 15 | 4333 | |||
XM_011536424.1 | XP_011534726.1 | 16 | 4458 | |||
GenBank mRNA | AK021747.1 | - | 1 | 1822 | ||
AK126521.1 | - | 12 | 3710 | |||
AK129995.1 | - | 1 | 2161 | |||
AL833158.1 | - | 15 | 4275 | |||
AL833380.1 | - | 9 | 3303 | |||
AL833661.1 | - | 9 | 3285 | |||
BC020521.2 | - | 14 | 4012 | 3914276 | ||
BX647239.1 | - | 5 | 4635 | |||
BX647863.1 | - | 14 | 3978 | |||
BX648295.1 | - | 5 | 4636 |