Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039355.2 | NP_001034444.1 | 4 | 2376 | ||
NM_001291813.1 | NP_001278742.1 | 5 | 2744 | |||
NM_001291814.1 | NP_001278743.1 | 5 | 2448 | |||
NM_152333.3 | NP_689546.1 | 3 | 2332 | |||
XM_006720038.1 | XP_006720101.1 | 6 | 2996 | |||
XM_006720039.3 | XP_006720102.1 | 5 | 2479 | |||
XM_011536443.1 | XP_011534745.1 | 7 | 3052 | |||
XM_011536444.1 | XP_011534746.1 | 5 | 2837 | |||
XM_011536446.1 | XP_011534748.1 | 5 | 3150 | |||
XM_011536447.1 | XP_011534749.1 | 4 | 3088 | |||
XM_011536448.1 | XP_011534750.1 | 4 | 3078 | |||
XM_017020979.1 | XP_016876468.1 | 5 | 2522 | |||
XM_017020980.1 | XP_016876469.1 | 5 | 7650 | |||
XM_017020981.1 | XP_016876470.1 | 4 | 7174 | |||
XM_017020982.1 | XP_016876471.1 | 4 | 5704 | |||
XM_017020983.1 | XP_016876472.1 | 4 | 5694 | |||
XM_017020984.1 | XP_016876473.1 | 4 | 5408 | |||
XM_017020985.1 | XP_016876474.1 | 5 | 5453 | |||
XM_017020986.1 | XP_016876475.1 | 4 | 4889 | |||
XM_017020987.1 | XP_016876476.1 | 5 | 4869 | |||
GenBank mRNA | AK095532.1 | - | 2 | 2816 | ||
AK096294.1 | - | 3 | 2224 | |||
AK123821.1 | - | 4 | 2268 | |||
AK172788.1 | - | 4 | 2298 | |||
AL832299.1 | - | 2 | 4871 | |||
BX247983.1 | - | 4 | 2352 |